Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702306

Sigma-Aldrich

Anti-HEC1/NDC80 Antibody, clone 4W2Y2, Rabbit Monoclonal

Synonym(s):

HEC, HEC1, HsHec1, KNTC2, TID3, hsNDC80

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₹40,548.30

₹40,548.30


Estimated to ship on27 March 2025Details



Select a Size

Change View
100 μL
₹40,548.30

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₹40,548.30


Estimated to ship on27 March 2025Details


biological source

rabbit

Quality Level

material

colorless

clone

4W2Y2, monoclonal

form

liquid

mol wt

76 kDa

species reactivity

human

concentration

0.16 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

ESTVNQGLSEAMNELDAVQREYQLVVQTTTEERRKVGNNLQRLLEMVATHVGSVEKHLEEQIAKVDREYEECMSEDLSENIKEIRDKYEKKATLIKSSEE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDC80(10403)

General description

This gene encodes a component of the NDC80 kinetochore complex. The encoded protein consists of an N-terminal microtubule binding domain and a C-terminal coiled-coiled domain that interacts with other components of the complex. This protein functions to organize and stabilize microtubule-kinetochore interactions and is required for proper chromosome segregation. [provided by RefSeq, Oct 2011]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 543-642 of human HEC1/NDC80 (O14777).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service