Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702278

Sigma-Aldrich

Anti-Annexin A11 Antibody, clone 8O5Z5, Rabbit Monoclonal

Synonym(s):

ANXA11,ANX11,CAP50

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₹40,548.30

₹40,548.30


Estimated to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
₹40,548.30

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₹40,548.30


Estimated to ship on27 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

8O5Z5, monoclonal

form

liquid

mol wt

56 kDa

species reactivity

mouse, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

FDIYEIKEAIKGVGTDEACLIEILASRSNEHIRELNRAYKAEFKKTLEEAIRSDTSGHFQRLLISLSQGNRDESTNVDMSLAQRDAQELYAAGENRLGTDESKFNAVLCSRSRAHLVAVFNEYQRMTGRDIEKSICREMSGDLEEGMLAVVKCLKNTPAFFAERLNKAMRGAGTKDRTLIRIMVSRSETDLLDIRSEYKRMYGKSLYHDISGDTSGDYRKILLKICGGND

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA11(311)

General description

This gene encodes a member of the annexin family, a group of calcium-dependent phospholipid-binding proteins. Annexins have unique N-terminal domains and conserved C-terminal domains, which contain calcium-dependent phospholipid-binding sites. The encoded protein is a 56-kD antigen recognized by sera from patients with various autoimmune diseases. Several transcript variants encoding two different isoforms have been identified.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 276-505 of human Annexin A11 (NP_001148.1).

Application

WB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service