Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702074

Sigma-Aldrich

Anti-ErbB4/HER4 Antibody, clone 8E5D0, Rabbit Monoclonal

Synonym(s):

ALS19,HER4,p180erbB4,ERBB4,HER4/ErbB4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₹40,548.30

₹40,548.30


Estimated to ship on08 May 2025Details



Select a Size

Change View
100 μL
₹40,548.30

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₹40,548.30


Estimated to ship on08 May 2025Details


biological source

rabbit

Quality Level

material

colorless

clone

8E5D0, monoclonal

form

liquid

mol wt

180 kDa

species reactivity

rat, mouse

concentration

0.25 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

DEYVNEPLYLNTFANTLGKAEYLKNNILSMPEKAKKAFDNPDYWNHSLPPRSTLQHPDYLQEYSTKYFYKQNGRIRPIVAENPEYLSEFSLKPGTVLPPPPYRHRNTVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ERBB4(2066)

General description

This gene is a member of the Tyr protein kinase family and the epidermal growth factor receptor subfamily. It encodes a single-pass type I membrane protein with multiple cysteine rich domains, a transmembrane domain, a tyrosine kinase domain, a phosphotidylinositol-3 kinase binding site and a PDZ domain binding motif. The protein binds to and is activated by neuregulins and other factors and induces a variety of cellular responses including mitogenesis and differentiation. Multiple proteolytic events allow for the release of a cytoplasmic fragment and an extracellular fragment. Mutations in this gene have been associated with cancer. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1200-1308 of human ErbB4/HER4 (Q15303).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service