Skip to Content
Merck
All Photos(1)

Key Documents

SAB4100213

Sigma-Aldrich

Monoclonal Anti-FAM127C antibody produced in mouse

clone FAM127C -A139, culture supernatant

Synonym(s):

AntiCXX1c, AntiFamily with sequence similarity 127, member C, AntiMAR8B, AntiOTTHUMP00000195547

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

FAM127C -A139, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 13.1 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

Gene Information

General description

FAM127C or retrotransposon Gag like 8B (RTL8B) is a member of the family with sequence similarity 127. FAM127C is localized on human chromosome Xq26.3.

Immunogen

FAM127C purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiedo Hidaka et al.
Oncotarget, 3(1), 44-57 (2012-02-02)
MicroRNAs (miRNA) are non-coding RNAs, approximately 22 nucleotides in length, which function as post-transcriptional regulators. A large body of evidence indicates that miRNAs regulate the expression of cancer-related genes involved in proliferation, migration, invasion, and metastasis. The aim of
C Selmi et al.
Clinical and experimental immunology, 169(3), 253-262 (2012-08-07)
Scleroderma (SSc) is a rare connective tissue disease characterized by fibrosis, microvasculopathy and autoimmune features. The role of genetics is limited in SSc, as suggested by similar concordance rates in monozygotic and dizygotic twin pairs, while environmental factors may act

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service