Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401270

Sigma-Aldrich

Monoclonal Anti-OCA2 antibody produced in mouse

clone 1A11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

BEY, BEY1, BEY2, BOCA, D15S12, EYCL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₹41,447.40

₹41,447.40


Estimated to ship on16 April 2025Details



Select a Size

Change View
100 μG
₹41,447.40

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

₹41,447.40


Estimated to ship on16 April 2025Details


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OCA2(4948)

General description

This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism. (provided by RefSeq)

Immunogen

OCA2 (AAH12097, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service