S5670
Sendai virus Nucleoprotein Fragment 321-336
≥90% (HPLC)
Synonym(s):
His-Gly-Glu-Phe-Ala-Pro-Gly-Asn-Tyr-Pro-Ala-Leu-Trp-Ser-Tyr-Ala
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C85H111N20O23
CAS Number:
Molecular Weight:
1780.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥90% (HPLC)
UniProt accession no.
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(C)NC(=O)C1CCCN1C(=O)C(Cc2ccc(O)cc2)NC(=O)C(CC(N)=O)NC(=O)CNC(=O)C3CCCN3C(=O)C(C)NC(=O)C(Cc4ccccc4)NC(=O)C(CCC(O)=O)NC(=O)CNC(=O)C(N)Cc5cnc[nH]5)C(=O)NC(Cc6c[nH]c7ccccc67)C(=O)NC(CO)C(=O)NC(Cc8ccc(O)cc8)C(=O)NC(C)C(O)=O
Gene Information
Sendai virus ... NP(1489779)
Biochem/physiol Actions
Peptide recognized by cytotoxic T lymphocytes in vivo. Mice immunized with fragment were protected from lethal virus infection.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
W M Kast et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(6), 2283-2287 (1991-03-15)
The only peptide of Sendai virus that is recognized by cytotoxic T lymphocytes (CTL) in B6 mice was found with (i) the use of recombinant vaccinia virus constructs containing separate genes of Sendai virus and (ii) a set of overlapping
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service