Skip to Content
Merck
All Photos(1)

Documents

R4282

Sigma-Aldrich

RF9 trifluoroacetate salt

≥97% (HPLC)

Synonym(s):

1-Adamantanecarbonyl-Arg-Phe-NH2 trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H38N6O3 · xC2HF3O2
CAS Number:
Molecular Weight:
482.62 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

solubility

H2O: 20 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.NC(=N)NCCC[C@H](NC(=O)C12C[C@H]3C[C@H](C[C@H](C3)C1)C2)C(=O)N[C@@H](Cc4ccccc4)C(N)=O

InChI

1S/C26H38N6O3.C2HF3O2/c27-22(33)21(12-16-5-2-1-3-6-16)31-23(34)20(7-4-8-30-25(28)29)32-24(35)26-13-17-9-18(14-26)11-19(10-17)15-26;3-2(4,5)1(6)7/h1-3,5-6,17-21H,4,7-15H2,(H2,27,33)(H,31,34)(H,32,35)(H4,28,29,30);(H,6,7)/t17-,18+,19-,20-,21-,26-;/m0./s1

InChI key

GPVFYPYIOXHDCU-CFIUKZCSSA-N

Biochem/physiol Actions

RF9 is a neuropeptide FF receptor antagonist. NPFF is a member of the FMRFamide (Phe-Met-Arg-PheNH2) family of neuropeptides, which are implicated in nociception and regulation of opioid analgesia. NPFF exhibits antiopioid activity (hyperalgesia). NPFF antagonists are expected to improve the therapeutic efficacy of opioids for treatment of chronic pain. The affinity of RF9 for NPFF1R and NPFF2R is 58 and 75 nM, respectively. RF9 acts as a potent competitive antagonist of NPFF at NPFF2R RF9 exhibits no agonist activity in GTPgS assays.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service