Skip to Content
Merck
All Photos(3)

Documents

R1282

Sigma-Aldrich

Anti-RNF149 (231-245) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DNAPTP2, Anti-Ring finger protein 149

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF149(284996)

Immunogen

synthetic peptide corresponding to amino acids 231-245 of human RNF149

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Xia et al.
Neuron, 85(5), 967-981 (2015-03-06)
Presenilins play essential roles in memory formation, synaptic function, and neuronal survival. Mutations in the Presenilin-1 (PSEN1) gene are the major cause of familial Alzheimer's disease (FAD). How PSEN1 mutations cause FAD is unclear, and pathogenic mechanisms based on gain
D J Watson et al.
The Biochemical journal, 340 ( Pt 3), 703-709 (1999-06-09)
Hereditary cerebral haemorrhage with amyloidosis, Dutch type (HCHWA-D), is a cerebral amyloidosis characterized by prominent vascular deposits and fatal haemorrhages. The disorder is caused by a point mutation in codon 693 of the gene encoding the amyloid precursor protein (APP)
Dan Frenkel et al.
The Journal of clinical investigation, 115(9), 2423-2433 (2005-08-16)
Amyloid beta-peptide (Abeta) appears to play a key pathogenic role in Alzheimer disease (AD). Immune therapy in mouse models of AD via Abeta immunization or passive administration of Abeta antibodies markedly reduces Abeta levels and reverses behavioral impairment. However, a
Eloise Hudry et al.
Science translational medicine, 5(212), 212ra161-212ra161 (2013-11-22)
Inheritance of the ε4 allele of apolipoprotein E (APOE) is the strongest genetic risk factor associated with the sporadic form of Alzheimer's disease (AD), whereas the rare APOE ε2 allele has the opposite effect. However, the mechanisms whereby APOE confers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service