Skip to Content
Merck
All Photos(1)

Key Documents

PZ0389

Sigma-Aldrich

PF-06869206

≥98% (HPLC)

Synonym(s):

(S)-3-Chloro-7-(2-(hydroxymethyl)morpholino)-2-methyl-5-(trifluoromethyl)-1H-pyrrolo[3,2-b]pyridine-6-carbonitrile, PF 06869206, PF06869206

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
₹7,112.03
25 MG
₹38,796.80

₹7,112.03


Estimated to ship on11 July 2025Details

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
₹7,112.03
25 MG
₹38,796.80

About This Item

Empirical Formula (Hill Notation):
C15H14ClF3N4O2
CAS Number:
Molecular Weight:
374.75
UNSPSC Code:
51111800
NACRES:
NA.77

₹7,112.03


Estimated to ship on11 July 2025Details

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=C(C)NC2=C1N=C(C(F)(F)F)C(C#N)=C2N3C[C@@H](CO)OCC3

InChI

1S/C15H14ClF3N4O2/c1-7-10(16)11-12(21-7)13(23-2-3-25-8(5-23)6-24)9(4-20)14(22-11)15(17,18)19/h8,21,24H,2-3,5-6H2,1H3/t8-/m0/s1

InChI key

ATFQBBCQZKVZJN-QMMMGPOBSA-N

Application

PF-06869206 has been used to selectively inhibit the activity of sodium-phosphate cotransporter 2a (NaPi2a).[1]

Biochem/physiol Actions

PF-06869206 is a selective inhibitor against sodium-phosphate cotransporter 2A (NaPi-2a, NaPi-Iia, Na+/Pi cotransporter 2A, Na+-dependent phosphate cotransporter 2A).
PF-06869206 may be used to treat patients with mineral and hormonal derangements linked with increased cardiovascular risk in chronic kidney disease. This drug might also improve urinary phosphate excretion.[2] PF-06869206 has an ability to decrease phosphate uptake in human embryonic kidney (HEK) cells transfected with mouse or rat renal sodium-dependent phosphate transporter genes (Npt2a).[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vivo effects of renal Npt2a inhibition
Thomas L, et al.
Faseb Journal, 33(1-supplement), 751-752 (2019)
Discovery of Orally Bioavailable Selective Inhibitors of the Sodium-Phosphate Cotransporter NaPi2a (SLC34A1)
Filipski KJ, et al.
ACS Medicinal Chemistry Letters, 9(5), 440-445 (2018)
Synthesis of PF-06869206
Kocienski P
Synfacts, 14(07), 0675-0675 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service