Skip to Content
Merck
All Photos(1)

Documents

PCG3007

Sigma-Aldrich

TruPAGE Running Antioxidant

800 ×

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NB.22

concentration

800 ×

compatibility

for use with Sigma-Aldrich TruPAGE Precast Gels
for use with Sigma-Aldrich TruPAGE SDS Running Buffers (PCG3001 and PCG3003)

storage temp.

2-8°C

Features and Benefits

TruPAGE Running Antioxidant is used with TruPAGE running buffers to maintain consistently high protein band resolution and sharpness. The antioxidant reagent reduces the gel environment to prevent potential sample reoxidation of reduced protein samples during electrophoresis. To obtain optimal results, supplement the inner chamber′s running buffer with TruPAGE Running Antioxidant prior to electrophoresis of reduced protein samples.

Legal Information

TruPAGE is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service