Skip to Content
Merck
All Photos(1)

Documents

P9978

Sigma-Aldrich

Anti-Potassium Channel Kir2.3 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:200-1:300

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KCNJ4(3761)
mouse ... Kcnj4(16520)
rat ... Kcnj4(116649)

Specificity

Directed against inward rectifying potassium channel Kir2.3 (IRK3, BIR11, Kcnj4).

Immunogen

highly purified synthetic peptide corresponding to amino acids 418-437 of rat Kir2.3. The epitope is identical in mouse and highly homologous in hamster, human, and Xenopus.

Physical form

Lyophilized at ~0.3 mg/ml from phosphate buffered saline, pH 7.4, 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service