Skip to Content
Merck
All Photos(1)

Key Documents

P1502

Sigma-Aldrich

PTH-(ε-phenylthiocarbamyl)lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4OS2
CAS Number:
Molecular Weight:
398.54
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCNC(=S)Nc2ccccc2)NC(=S)N1c3ccccc3

InChI

1S/C20H22N4OS2/c25-18-17(23-20(27)24(18)16-11-5-2-6-12-16)13-7-8-14-21-19(26)22-15-9-3-1-4-10-15/h1-6,9-12,17H,7-8,13-14H2,(H,23,27)(H2,21,22,26)/t17-/m0/s1

InChI key

BUPYRERCBPFRFM-KRWDZBQOSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service