P1201
DL-Propranolol-[4-3H] hydrochloride
ethanol solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H21NO2 · HCl
CAS Number:
Molecular Weight:
295.80
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form
ethanol solution
extent of labeling
15-30 Ci/mmol
mass shift
M+2
storage temp.
2-8°C
SMILES string
Cl.[3H]c1ccc(OCC(O)CNC(C)C)c2ccccc12
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
application for HPLCapplication for SFCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service