Skip to Content
Merck
All Photos(1)

Key Documents

P0257

Sigma-Aldrich

D(+)-2-Phosphoglyceric acid sodium salt hydrate

Synonym(s):

D-Glycerate 2-phosphate sodium salt, Sodium D-2-phosphoglycerate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7O7P · xNa+ · yH2O
CAS Number:
Molecular Weight:
186.06 (anhydrous free acid basis)
UNSPSC Code:
12352200

concentration

≥50% (enzymatic)

storage temp.

−20°C

SMILES string

O=C(O)[C@@H](CO)OP(O)(O)=O.C.C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Enantiomerically pure metabolite of glycolysis/gluconeogenesis and also substrate for a number of important enzymes in central metabolism like enolase and phosphoglycerate mutase.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service