Skip to Content
Merck
All Photos(1)

Key Documents

P0086

Sigma-Aldrich

Protargol S

certified by the Biological Stain Commission

Synonym(s):

Silver proteinate, Albumose silver, Protargol, Silver protein, strong

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

certified by the Biological Stain Commission

Looking for similar products? Visit Product Comparison Guide

Suitability

Silver albumose intended for staining nerve tissue by a modified Bodian technique.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service