Key Documents
OH30073979
Ohaus® Pen pH Meter
Model ST10T-B
Select a Size
1 EACH
₹12,100.00
₹12,100.00
Estimated to ship on27 May 2025Details
Select a Size
Change View
1 EACH
₹12,100.00
About This Item
UNSPSC Code:
41115603
₹12,100.00
Estimated to ship on27 May 2025Details
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30073979
TDS measuring range
0.0-1000.0 mg/L
battery power
4 × 1.5 V, 200 hr life
General description
With durable IP67 waterproof ABS housing, protective sensor cap, and an automatic shutdown feature that preserves battery life, OHAUS pen meters can endure consistent use in rough, wet environments.
Application
pH, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), salinity, total dissolved solids (TDS), temperature measurement
Features and Benefits
Easily replaceable electrodes, automatic shut off feature
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)