Skip to Content
Merck
All Photos(1)

Key Documents

O0375

Sigma-Aldrich

Octylamine

Synonym(s):

1-Aminooctane, n-Octylamine, Caprylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7NH2
CAS Number:
Molecular Weight:
129.24
Beilstein:
1679227
EC Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 20 °C)

refractive index

n20/D 1.429 (lit.)

bp

175-177 °C (lit.)

mp

−5-−1 °C (lit.)

density

0.782 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN

InChI

1S/C8H19N/c1-2-3-4-5-6-7-8-9/h2-9H2,1H3

InChI key

IOQPZZOEVPZRBK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosario Zamora et al.
Journal of agricultural and food chemistry, 54(6), 2398-2404 (2006-03-16)
The reactions of 4,5-epoxy-2-decenal with octylamine, benzylamine, and 2-phenylglycine methyl ester were studied to investigate if amines may suffer a Strecker type degradation by epoxyalkenals analogously to amino acids. In addition to other reactions, the studied amines were converted into
Vera Bocharova et al.
Small (Weinheim an der Bergstrasse, Germany), 2(7), 910-916 (2006-12-29)
Spin-coating of isolated positively charged macromolecules onto mica in the presence of octylamine was found to be a simple and general method of stretching and aligning the macromolecular chains. The contour length and molar mass for the stretched macromolecules can
Brad A Kairdolf et al.
Analytical chemistry, 80(8), 3029-3034 (2008-03-08)
Quantum-dot (QD) nanocrystals are promising fluorescent probes for multiplexed staining assays in biological applications. However, nonspecific QD binding to cellular membranes and proteins remains a limiting factor in detection sensitivity and specificity. Here we report a new class of hydroxyl
Hong Zheng et al.
Journal of biomedical nanotechnology, 7(5), 648-658 (2011-12-27)
Quantum dots (QDs) have emerged as alternative or complementary tools to organic fluorescent dyes currently used in bioimaging. QDs hold several advantages over conventional fluorescent dyes including greater photostability and a wider range of excitation/emission wavelengths. However, recent work suggests
Shibin He et al.
Biomaterials, 32(23), 5471-5477 (2011-05-07)
Quantum dots (QDs) have been received most attention due to their unique properties. Constructing QDs conjugated with certain number of biomolecules is considered as one of the most important research goals in nanobiotechnology. In this study, we report polymerase chain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service