Skip to Content
Merck
All Photos(2)

Key Documents

N5762

Sigma-Aldrich

3-(2-Naphthyl)-DL-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO2
CAS Number:
Molecular Weight:
215.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

form

powder

storage temp.

2-8°C

SMILES string

NC(Cc1ccc2ccccc2c1)C(O)=O

InChI

1S/C13H13NO2/c14-12(13(15)16)8-9-5-6-10-3-1-2-4-11(10)7-9/h1-7,12H,8,14H2,(H,15,16)

InChI key

JPZXHKDZASGCLU-UHFFFAOYSA-N

Biochem/physiol Actions

3-(2-Naphthyl)-DL-alanine is an unnatural amino acid. The L-enantiomer has been incorporated into E. coli showing allowing for further understanding of protein function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang et al.
Journal of the American Chemical Society, 124(9), 1836-1837 (2002-02-28)
An unnatural amino acid, L-3-(2-naphthyl)alanine, has been site-specifically incorporated into proteins in Escherichia coli. An orthogonal aminoacyl-tRNA synthetase was evolved that uniquely aminoacylates the unnatural amino acid onto an orthogonal amber suppressor tRNA, which delivers the acylated amino acid in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service