Skip to Content
Merck
All Photos(1)

Key Documents

N3637

Sigma-Aldrich

Nle-Arg-Phe amide

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H35N7O3
CAS Number:
Molecular Weight:
433.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CCCC[C@H](N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc1ccccc1)C(N)=O

InChI

1S/C21H35N7O3/c1-2-3-10-15(22)19(30)27-16(11-7-12-26-21(24)25)20(31)28-17(18(23)29)13-14-8-5-4-6-9-14/h4-6,8-9,15-17H,2-3,7,10-13,22H2,1H3,(H2,23,29)(H,27,30)(H,28,31)(H4,24,25,26)/t15-,16-,17-/m0/s1

InChI key

XPMYDYSYRCLZEG-ULQDDVLXSA-N

Biochem/physiol Actions

Nle-Arg-Phe amide is a molluscan cardioexcitatory neuropeptide analog.

Linkage

FMRF amide analog; molluscan cardioexcitatory neuropeptide analog

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of peptide-like substances in the Placopecten: Possible role in growth and reproduction
Pani, A.K.; Roghani, A.; and Croll, R.P.
Biogenic Amines, 19, 47-67 (2005)
Caroline Perner et al.
Immunity, 53(5), 1063-1077 (2020-10-26)
Dendritic cells (DCs) of the cDC2 lineage initiate allergic immunity and in the dermis are marked by their expression of CD301b. CD301b+ dermal DCs respond to allergens encountered in vivo, but not in vitro. This suggests that another cell in the dermis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service