Skip to Content
Merck
All Photos(1)

Documents

MSST0016

Sigma-Aldrich

SILuLite B2M beta-2-microglobulin human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

β-2-microglobulin, Mass spectrometry standard, B@M, Mass spectrometry standard, beta-2-microglobulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥98% (SDS-PAGE)

form

lyophilized powder

technique(s)

mass spectrometry (MS): suitable

suitability

suitable for mass spectrometry (internal calibrator)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... B2M(567)

General description

SILu Lite B2M is a recombinant human protein expressed in human 293 cells. It is a monomer of 119 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~14 kDa. SILu Lite B2M is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.
Suggested Quantitative Analysis Parameters
(MRM settings provided for three suggested peptides)

Biochem/physiol Actions

B2M is the light chain of the major histocompatibility class (MHC) I molecule expressed on the cell surface of all nucleated cells. Increased urinary B2M excretion has been observed to be an early marker of tubular injury in a number of settings, including nephrotoxicant exposure, cardiac surgery, and renal transplantation, preceding rises in serum creatinine by as many as 4−5 days. B2M may also serve as an early biomarker for AKI (Acute Kidney Injury).

Sequence

IQRTPKIQVYSRHPAENGKSNFLNCYVSGFHPSDIEVDLLKNGERIEKVEHSDLSFSKDWSFYLLYYTEFTPTEKDEYACRVNHVTLSQPKIVKWDRDMDYKDDDDKGHHHHHHHHGGQ

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), licensing@dupont.com.
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service