Skip to Content
Merck
All Photos(1)

Key Documents

MAK410

Sigma-Aldrich

Urea Colorimetric Assay Kit II

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

detection method

colorimetric

relevant disease(s)

orthopedic diseases; genitourinary disorders

storage temp.

2-8°C

General description

The new Urea Assay Kit, MAK471, now available!
Urea is a waste product which is produced in the liver, dissolved and transported in the blood (in a concentration of 1.6-7.5 mM), and excreted by the kidney. Urea plays a very important role in protein catabolism, removal of toxic ammonia from the body, and the countercurrent system, which allows for reabsorption of water and critical ions in the nephrons. Urea concentration is an important indicator for assessing the function of the kidneys and other organs.

Features and Benefits

Urea Colorimetric Assay Kit II:
  • Simple procedure with results in ∼1hour.
  • Sensitive, detecting less than 10 μM of urea.

Suitability

The kit is suitable for the measurement of urea levels in biological samples (urine, serum, plasma etc.), tissue (liver, kidney etc.), adherent or suspension cells (3T3 cells, Jurkat cells etc.) and milk. It is also suitable for the analysis of liver and kidney function.

Principle

The Urea Colorimetric Assay Kit II is based on Jung′s method with a modification that delivers more robust and sensitive data. The urea is condensed with o-phthalaldehyde (OPA), followed by a reaction to form a colored product with strong absorbance at A505 nm.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service