We do not have data supporting frozen samples and recommend using fresh samples. However, provided this is not available, it would be prudent to compare the readings from a sample frozen versus fresh to see if there is any impact on the determination.
MAK357
Hydroxyproline Assay Kit (Perchlorate-Free)
sufficient for 100 colorimetric tests
About This Item
Recommended Products
usage
sufficient for 100 colorimetric tests
detection method
colorimetric
relevant disease(s)
rheumatological diseases; cancer; neurological disorders; cardiovascular diseases
storage temp.
−20°C
General description
Features and Benefits
Suitability
Principle
replaced by
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
188.6 °F
Flash Point(C)
87 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hi, I was wondering if the tissue has to be fresh before homogenization and if freeze and thaw would affect the accuracy of the assay
1 answer-
Helpful?
-
-
Can Product MAK357, Hydroxyproline Assay Kit Perchlorate-Free, be used to measure the hydroxyproline levels in cell culture supernatants?
1 answer-
The kit is optimized for use in determination of hydroxyproline in biological samples (e.g. plasma, serum, cell lysate, tissue samples.) This kit has not been tested for use with cell culture supernatant. The presence of phenol red may interfere with results. In addition, the cultured cell line may not secrete sufficient hydroxyproline to be detected. It would be up to the end user to determine suitability.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service