Skip to Content
Merck
All Photos(2)

Documents

MAK191

Sigma-Aldrich

Carrez Clarification Reagent Kit

Sufficient for 100 tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

sufficient for 100 tests

storage temp.

room temp

General description

The Carrez Clarification Reagent Kit is suitable for the preparation of samples for the analysis of small molecules such as carbohydrates, alcohols, aldehydes, and organic acids. The samples might contain proteins, fats, and redox compounds that interfere with the analysis.

Application

The Carrez Clarification Reagent Kit can be used for the preparation of food (such as meat, cereal, or milk) or blood for further analysis (such as HPLC or bioassays). The reagents in this kit cannot be used to prepare samples that contain or may be converted to ascorbate, ammonia, or citrate in enzyme-based assays.

Suitability

Suitable for the clarification of food, beverage, and other samples for use in downstream analyses.

Principle

The Carrez Clarification Reagents remove interfering compounds from the analytes and are more simple, less expensive, and less time-consuming than using spin filters.

Kit Components Only

Product No.
Description

  • Carrez Reagent I

  • Carrez Reagent II

replaced by

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of convective drying pretreatment on the physicochemical properties of black garlic.
Rios-Rios KL, et al.
Acta horticulturae, 1194, 845-852 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service