Skip to Content
Merck
All Photos(1)

Documents

M8563

Sigma-Aldrich

Mimetic Red 3 ALPHA Column

suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

description

10 ml

Quality Level

form

suspension

shipped in

wet ice

storage temp.

2-8°C

Application

Mimetic columns are used in affinity chromatography and protein chromatography. Mimetic columns have been used in studies to develop anti-inflammatory therapeutics.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Clark et al.
FEBS letters, 471(2-3), 182-186 (2000-04-18)
Integrin alpha4beta1 is a major leukocyte adhesion receptor that is a key target for the development of anti-inflammatory therapeutics. With the dual long-term goals of developing a reagent for use in high-throughput inhibitor screening assays and for crystallisation trials and
Tsutomu Arakawa et al.
Protein expression and purification, 63(2), 158-163 (2008-10-14)
MEP HyperCel resin, one of the Protein-A mimetic columns, is designed to bind antibodies at physiological pH and elutes the bound antibodies at mildly acidic pH. We have tested aqueous arginine solution for washing and elution of the resin. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service