Skip to Content
Merck
All Photos(1)

Key Documents

M6405

Sigma-Aldrich

2-O-Methyl-α-D-N-acetylneuraminic acid

~95% (TLC)

Synonym(s):

α-D-Neu-5-Ac-2→OCH3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO9
CAS Number:
Molecular Weight:
323.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
To inquire about this product M6405, please contact your local Merck office or dealer. Contact Technical Service

Assay

~95% (TLC)

storage temp.

−20°C

SMILES string

COC1(CC(O)C(NC(C)=O)C(O1)C(O)C(O)CO)C(O)=O

InChI

1S/C12H21NO9/c1-5(15)13-8-6(16)3-12(21-2,11(19)20)22-10(8)9(18)7(17)4-14/h6-10,14,16-18H,3-4H2,1-2H3,(H,13,15)(H,19,20)

InChI key

NJRVVFURCKKXOD-UHFFFAOYSA-N

General description

N-acetylneuraminic acid is the minimum binding epitope of rotavirus to host cells.

Application

2-O-Methyl-α-D-N-acetylneuraminic acid is a model compound for studies assessing the binding of influenza virus hemagglutinin[1] and metal ions.[2] N-acetylneuraminic acid has been used in a study to assess lactose-based sialylmimetics inhibition of the rhesus and human rotavirus strains. [3] It has also been used in a study to investigate the binding specificity of botulinum C16S toxin HA1 to various sugars. [4]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N K Sauter et al.
Biochemistry, 28(21), 8388-8396 (1989-10-17)
The equilibrium binding of influenza virus hemagglutinin to derivatives of its cell-surface ligand, sialic acid, was measured by nuclear magnetic resonance (NMR) spectroscopy. Binding was quantified by observing perturbations of sialic acid resonances in the presence of protein. The major
Toshio Nakamura et al.
Journal of molecular biology, 376(3), 854-867 (2008-01-08)
Clostridium botulinum type C 16S progenitor toxin contains a hemagglutinin (HA) subcomponent, designated HA1, which appears to play an important role in the effective internalization of the toxin in gastrointestinal epithelial cells and in creating a broad specificity for the
Angela Liakatos et al.
Bioorganic & medicinal chemistry, 14(3), 739-757 (2005-10-11)
Rotaviruses are the most significant cause of gastroenteritis in young children and are responsible for over 600,000 infant deaths annually. The rotaviral haemagglutinin protein (VP8*) of some strains has been implicated in early recognition and binding events of host cell-surface
Daman, M.E., and Dill, K.
Carbohydrate Research, 102, 47-47 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service