Skip to Content
Merck
All Photos(1)

Key Documents

M5153

Sigma-Aldrich

8-Methylnonanoic acid

≥97% (capillary GC), ≥97% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2
CAS Number:
Molecular Weight:
172.26
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97% (capillary GC)
≥97% (titration)

storage temp.

−20°C

SMILES string

CC(C)CCCCCCC(O)=O

Looking for similar products? Visit Product Comparison Guide

Application

Intermediate in synthesis of dihydrocapsaicin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jones, S., et al.
J. Chem. Res. Synop., 5, 344-344 (1987)
Janclei P Coutinho et al.
Talanta, 134, 256-263 (2015-01-27)
In this work a multivariate statistical tool (Derringer and Suich optimization) was proposed for the separation of seventeen capsinoids (natural and synthetic) using the UHPLC-DAD chromatography. Capsinoids were analyzed at 280 nm. The variables optimized were the mobile phase (water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service