Skip to Content
Merck
All Photos(1)

Key Documents

M2640

SAFC

Methoxyacetyl chloride

Synonym(s):

Methoxyacetic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2COCl
CAS Number:
Molecular Weight:
108.52
Beilstein:
1740244
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

refractive index

n20/D 1.419 (lit.)

bp

112-113 °C (lit.)

density

1.187 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COCC(Cl)=O

InChI

1S/C3H5ClO2/c1-6-2-3(4)5/h2H2,1H3

InChI key

JJKWHOSQTYYFAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F

Flash Point(C)

28 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service