M204
p-MPPI monohydrochloride
solid, ≥98% (HPLC)
Synonym(s):
4-Iodo-N-[2-[4-(methoxyphenyl)-1-piperazinyl]ethyl]-N-(2-pyridinyl)benzamide monohydrochloride
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
solid
color
white
solubility
H2O: 2.4 mg/mL
SMILES string
Cl.COc1ccccc1N2CCN(CC2)CCN(C(=O)c3ccc(I)cc3)c4ccccn4
Gene Information
human ... HTR1A(3350)
Biochem/physiol Actions
Caution
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service