Skip to Content
Merck
All Photos(1)

Key Documents

L8016

Sigma-Aldrich

Anti-β-Lipoprotein (LDL) antibody produced in goat

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

goat

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

immunoelectrophoresis: 1:16

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOB(338)

General description

β-lipoprotein (ApoB) is a primary cholesterol carrying apolipoprotein of chylomicrons and low-density lipoproteins (LDL). ApoB is essential for LDL formation and LDL receptor binding. ApoB is involved in the delivery of cholesterol to cells.

Specificity

Goat polyclonal anti-β-Lipoprotein (LDL) antiserum is determined to be immunospecific for β-lipoprotein by immunoelectrophoresis against normal human serum. No reactivity is observed against α-lipoprotein (HDL, High Density Lipoprotein). Using immunoelectophoresis, a 1:16 dilution of Goat Anti-Human β-Lipoprotein shows strong reactivity with human β-lipoprotein at 1 mg/ml.

Application

Goat polyclonal anti-β-Lipoprotein (LDL) antibody is used to tag β-lipoprotein for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as immunoblotting, immunoprecipitation, and immunofluorescence. It is used as a probe to determine the presence and roles of β-lipoprotein in chylomicron and LDL formation and function especially involving cholesterol transport and deliver to cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service