Skip to Content
Merck
All Photos(1)

Key Documents

L3526

Sigma-Aldrich

Leukotriene A3 methyl ester

~50 μg/mL in hexane: diethyl ether: triethylamine (95:5:0.5), ≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O3
CAS Number:
Molecular Weight:
334.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

concentration

~50 μg/mL in hexane: diethyl ether: triethylamine (95:5:0.5)

functional group

ester

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCC/C=C\C=C\C=C\[C@@H]1O[C@H]1CCCC(=O)OC

InChI

1S/C21H34O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-16-19-20(24-19)17-15-18-21(22)23-2/h10-14,16,19-20H,3-9,15,17-18H2,1-2H3/b11-10-,13-12+,16-14+/t19-,20-/m0/s1

InChI key

YOAQIJHBMIIBJM-NLAMMTFYSA-N

Packaging

Packaged under Argon.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service