Skip to Content
Merck
All Photos(1)

Key Documents

L3031

Sigma-Aldrich

Lesquerolic acid methyl ester

Synonym(s):

(R)-(+)-14-Hydroxy-cis-11-eicosenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H40O3
CAS Number:
Molecular Weight:
340.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
Pricing and availability is not currently available.

storage temp.

−20°C

SMILES string

CCCCCC[C@@H](O)C\C=C/CCCCCCCCCC(=O)OC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Yeong Jeon et al.
Applied microbiology and biotechnology, 99(15), 6267-6275 (2015-02-01)
Enzyme fusion was investigated as a strategy to improve productivity of a two-step whole-cell biocatalysis. The biotransformation of long-chain sec-alcohols into esters by an alcohol dehydrogenase (ADH) and Baeyer-Villiger monooxygenases (BVMOs) was used as the model reaction. The recombinant Escherichia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service