Skip to Content
Merck
All Photos(1)

Key Documents

L1658

Sigma-Aldrich

Locustapyrokinin from Locusta migratoria

≥97% (HPLC)

Synonym(s):

pGlu-Asp-Ser-Gly-Asp-Glu-Trp-Pro-Gln-Gln-Pro-Phe-Val-Pro-Arg-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C85H121N23O26
CAS Number:
Molecular Weight:
1881.01
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@@H]3CCCN3C(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]4CCCN4C(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H]7CCC(=O)N7)C(C)C)C(N)=O

InChI

1S/C85H121N23O26/c1-42(2)34-53(70(88)120)100-72(122)48(18-10-30-91-85(89)90)97-80(130)61-21-13-33-108(61)84(134)69(43(3)4)105-78(128)54(35-44-14-6-5-7-15-44)102-81(131)60-20-11-31-106(60)82(132)52(23-27-63(87)111)99-74(124)50(22-26-62(86)110)98-79(129)59-19-12-32-107(59)83(133)57(36-45-39-92-47-17-9-8-16-46(45)47)103-75(125)51(25-29-66(114)115)96-76(126)55(37-67(116)117)95-65(113)40-93-71(121)58(41-109)104-77(127)56(38-68(118)119)101-73(123)49-24-28-64(112)94-49/h5-9,14-17,39,42-43,48-61,69,92,109H,10-13,18-38,40-41H2,1-4H3,(H2,86,110)(H2,87,111)(H2,88,120)(H,93,121)(H,94,112)(H,95,113)(H,96,126)(H,97,130)(H,98,129)(H,99,124)(H,100,122)(H,101,123)(H,102,131)(H,103,125)(H,104,127)(H,105,128)(H,114,115)(H,116,117)(H,118,119)(H4,89,90,91)/t48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,69-/m0/s1

InChI key

YDWOSJDFVOFKIC-ICOVXKSRSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Schoofs et al.
General and comparative endocrinology, 81(1), 97-104 (1991-01-01)
A neuropeptide which stimulates the motility of the cockroach hindgut has been isolated from an extract of 9000 brain-corpora cardiaca-corpora allata-subesophageal ganglion complexes of Locusta migratoria. Biological activity was monitored during HPLC purification by observing the myotropic effect of column
Cross-activity between pheromone biosynthesis activating neuropeptide (PBAN) and myotropic pyrokinin insect peptides.
H Kuniyoshi et al.
Bioscience, biotechnology, and biochemistry, 56(1), 167-168 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service