Skip to Content
Merck
All Photos(1)

Key Documents

J4205

Sigma-Aldrich

JK184

≥98% (HPLC), powder

Synonym(s):

N-(4-ethoxyphenyl)-4-(2-methylimidazo[1,2-a]pyridin-3-yl)-2-thiazolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18N4OS
CAS Number:
Molecular Weight:
350.44
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light brown

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

CCOc1ccc(Cc2nc(cs2)-c3c(C)nc4ccccn34)cc1

InChI

1S/C20H19N3OS/c1-3-24-16-9-7-15(8-10-16)12-19-22-17(13-25-19)20-14(2)21-18-6-4-5-11-23(18)20/h4-11,13H,3,12H2,1-2H3

InChI key

CBKFVRMNQUKWPX-UHFFFAOYSA-N

Biochem/physiol Actions

JK184 is a Hedgehog signaling pathway antagonist. JK184 blocks sonic hedgehog induced Gli transcription with an IC50 of 30 nM, and disrupts hedgehog signalling by inhibition of alcohol dehydrogenase 7 and depolymerization of mirotubules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen O Oladapo et al.
Cancer letters, 411, 136-149 (2017-10-03)
Activation of the Hedgehog (Hh) pathway effector GLI1 is linked to tumorigenesis and invasiveness in a number of cancers, with targeting of GLI1 by small molecule antagonists shown to be effective. We profiled a collection of GLI antagonists possessing distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service