IST111078SS
ClearASeal Perf™ heat sealable film
LabSheet™, W × L 78 mm × 125 mm, sterile, pack of 100 sheets
Synonym(s):
gas permeable seal, plate sealing film, sample incubation seal
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
sterility
sterile
packaging
pack of 100 sheets
manufacturer/tradename
IST Scientific IST-111-078SS
technique(s)
PCR: suitable
W × L
78 mm × 125 mm
Looking for similar products? Visit Product Comparison Guide
General description
Optically clear, gas permeable heat seal film, with 3 mm perforated slits for gas transfer. These slits render the seal gas permeable, whilst retaining evaporation to a minimum, compared to the use of lids.
Application
- Cell and Tissue culture
- Insect and seed storage as it enables gas exchange, whilst providing an inert surface with no adhesive to interfere with the well contents
- Short term incubation
Features and Benefits
- Seals polypropylene, polyethylene, polystyrene and cyclic olefin copolymer (COC) plates
- Seal integrity range: -80 °C to 110 °C
Legal Information
ClearASeal Peel is a trademark of IST Scientific
LabSheet is a trademark of IST Scientific
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service