Skip to Content
Merck
All Photos(1)

Key Documents

I9504

Sigma-Aldrich

Iodomethane-d3

Synonym(s):

Methyl-d3 Iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3I
CAS Number:
Molecular Weight:
144.96
Beilstein:
1732026
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

4.89 (vs air)

refractive index

n20/D 1.5262 (lit.)

bp

42 °C (lit.)

mp

−66.5 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])I

InChI

1S/CH3I/c1-2/h1H3/i1D3

InChI key

INQOMBQAUSQDDS-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service