Skip to Content
Merck
All Photos(1)

Documents

I6632

Sigma-Aldrich

N2-Isobutyryl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyguanosine

~98%

Synonym(s):

iBu-DMT-dG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H37N5O7
CAS Number:
Molecular Weight:
639.70
Beilstein:
3582365
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~98%

form

powder

mp

~150 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2O)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C35H37N5O7/c1-21(2)32(42)38-34-37-31-30(33(43)39-34)36-20-40(31)29-18-27(41)28(47-29)19-46-35(22-8-6-5-7-9-22,23-10-14-25(44-3)15-11-23)24-12-16-26(45-4)17-13-24/h5-17,20-21,27-29,41H,18-19H2,1-4H3,(H2,37,38,39,42,43)/t27-,28+,29+/m0/s1

InChI key

RMQXDNUKLIDXOS-ZGIBFIJWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service