Skip to Content
Merck
All Photos(1)

Key Documents

H9027

Sigma-Aldrich

8Z,11Z-Heptadecadienyl bromide

~250 mg/mL in diethyl ether, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥98%

concentration

~250 mg/mL in diethyl ether

storage temp.

−20°C

Application

Synthetic precursor to linoleic acid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service