Skip to Content
Merck
All Photos(1)

Key Documents

H6752

Sigma-Aldrich

8-Quinolinol hemisulfate salt

crystalline

Synonym(s):

8-Hydroxyquinoline hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O2 · H2SO4
CAS Number:
Molecular Weight:
388.39
Beilstein:
3760550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

description

C9H7NO · ½H2SO4 (Linear Formula)

form

crystalline

color

light yellow

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

OS(O)(=O)=O.Oc1cccc2cccnc12.Oc3cccc4cccnc34

InChI

1S/2C9H7NO.H2O4S/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;1-5(2,3)4/h2*1-6,11H;(H2,1,2,3,4)

InChI key

YYVFXSYQSOZCOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service