Skip to Content
Merck
All Photos(1)

Key Documents

H6639

Sigma-Aldrich

Methyl heptacosanoate

≥99%

Synonym(s):

Heptacosanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)25COOCH3
CAS Number:
Molecular Weight:
424.74
Beilstein:
1801611
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

form

powder

functional group

ester

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C28H56O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28(29)30-2/h3-27H2,1-2H3

InChI key

SCVPLVIKFSVDDH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Paik et al.
Journal of chromatography. B, Biomedical sciences and applications, 721(1), 3-11 (1999-02-23)
A sensitive method for very-long-chain fatty acid analysis was developed by gas chromatography-nitrogen-phosphorus detection by using cyanomethyl derivatization. Bromoacetonitrile as alkylating reagent was used to improve nitrogen phosphorus detection detectability of compounds containing non-nitrogen. The carboxyl group of very-long-chain fatty
R Aliya et al.
Pakistan journal of pharmaceutical sciences, 4(2), 103-111 (1991-07-01)
The green seaweed Codium iyengarii Bphirg., collected from the coast of Karachi, has been studied for its fatty acid composition. The acids were converted into methyl caters and identified by GC-MS, which revealed the presence of 11 saturated, 10 monoethenoic
Characterization and carbon metabolism in fungi pathogenic to Triatoma infestans, a chagas disease vector.
Juarez MP, Crespo R, Fernandez GC, et al.
J. Invert. Path., 76, 198-207 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service