H3021
Antiflammin-2
≥97% (HPLC)
Synonym(s):
Anti-inflammatory peptide 2, His-Asp-Met-Asn-Lys-Val-Leu-Asp-Leu
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C46H77N13O15S
CAS Number:
Molecular Weight:
1084.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥97% (HPLC)
composition
Peptide content, ~65%
storage temp.
−20°C
SMILES string
CSCCC(NC(=O)C(CC(O)=O)NC(=O)C(N)Cc1cnc[nH]1)C(=O)NC(CC(N)=O)C(=O)NC(CCCCN)C(=O)NC(C(C)C)C(=O)NC(CC(C)C)C(=O)NC(CC(O)=O)C(=O)NC(CC(C)C)C(O)=O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wayne B Anderson et al.
Chemical research in toxicology, 17(5), 650-662 (2004-05-18)
Dichloroacetic acid (DCA) is a drinking water contaminant, a therapeutic agent, and a rodent carcinogen. Glutathione transferase zeta (GSTZ1-1) catalyzes the biotransformation of a range of alpha-haloalkanoates and the cis-trans isomerization of maleylacetoacetate. GSTZ1-1 catalyzes the bioactivation of fluorine-lacking dihaloacetates
J M Ye et al.
Journal of pharmaceutical sciences, 85(7), 695-699 (1996-07-01)
Antiflammin 2 (HDMNKVLDL, AF2) is a synthetic peptide derived from the region of highest sequence similarity of lipocortin I and uteroglobin, and is a potent antiinflammatory agent without any known side effects of corticosteroids. The antiinflammatory activity of AF2 has
Degradation of antiflammin 2 in aqueous solution.
J L Wolfe et al.
Journal of pharmaceutical sciences, 83(12), 1762-1764 (1994-12-01)
S Lloret et al.
Biochemical pharmacology, 50(3), 347-353 (1995-07-31)
Antiflammins are synthetic peptides with sequence homology to proteins inhibitory for phospholipase A2 (EC 3.1.1.4). The effect of antiflammin 2 on murine arachidonate or 12-O-tetradecanoylphorbol 13-acetate (TPA)-induced ear oedema has been studied. Topical application of arachidonic acid (AA) produced a
J M Ye et al.
Pharmaceutical research, 13(2), 250-255 (1996-02-01)
To study the oxidation of the methionine residue of antiflammin 2 (HDMNKVLDL, AF2) as a function of pH, buffer concentration, ionic strength, and temperature using different concentrations of hydrogen peroxide and to determine the accessibility of methionine residue to oxidation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service