Key Documents
GN664160
Greiner CELLSTAR® dish
100 x 20 mm, 58 cm2, TC treated, sterile, w/ vents
Synonym(s):
cell culture dish, cell culture plate, cell culture plates, Greiner CELLSTAR® cell culture dishes
About This Item
UNSPSC Code:
41121812
Pricing and availability is not currently available.
packaging
pack of 360 ea
manufacturer/tradename
Greiner 664160
availability
available only in China
General description
The CELLSTAR®culture dish combines the convenience of astandard size 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high-resolution microscopic images of invitro cultivated cultures. Made from high-grade polystyrene combined with an integrated glass bottom. The innovative design of the dish provides a single-plane, flat bottom with a consistent working distance and maximal planarity. Moreover, the dish bottom configuration facilitates optimal thermal conductivity andavoids thermal variations in heated platforms used for live cell imaging.
Features and Benefits
  • Greiner CELLSTAR™ dishes are manufactured from highly transparent crystal clear polystyrene for optimal microscopic examinations.
  • Surface treated for optimal cell attachment and maximum cell proliferation.
  • Vented - to facilitate optimal gas exchange.
  • Dishes are stackable and provide a convenient space-saving option.
  • Free from detectable RNA, DNA, endotoxins, and human DNA.
  • Pyrogen-free and non-cytotoxic.
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)