Documents
GE28-4064-07
Sigma-Aldrich
Tricorn 10 Adapter Unit
About This Item
UNSPSC Code:
41115716
NACRES:
SB.52
technique(s)
LPLC: suitable
Features and Benefits
Glass tubes coated with plastic film protects individuals in the laboratory from harm if the column is dropped or operated outside its specification.
The columns are available with inner diameters of 5 and 10 mm, and in lengths from 20 to 200 mm. Column lengths of 300 and 600 mm are also available for the 10 mm diameter columns. The design and dimensions of the columns make them particularly useful for screening and polishing steps in protein purification.
Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures (see Specifications). They are suitable for use with high performance media, such as Sepharose High Performance, Superdex Prep Grade, and SOURCE.