Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1150-01

Silver Staining Kit, Protein

Cytiva 17-1150-01

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NA.32

feature

wetted part: no

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva 17-1150-01

storage temp.

room temp

General description

PlusOne Silver Staining Kit, Protein and PlusOne DNA Silver Staining Kit are two silver staining kits that bring speed, convenience, and very high sensitivity to the staining of proteins and nucleic acids in polyacrylamide electrophoresis. gels.

Silver Staining Kit, Protein includes detailed protocols and components in quantities sufficient to stain 10 DALT precast gels, 10 precast 12.5 x 26 cm gels, 20 mini slab gels (8–10 x 8 cm), or 30 to 60 PhastGels. It can also be used with the Processor Plus.

Application

For use in Hoefer automated gel stainer as well as for manual silver staining.

Features and Benefits

  • Based on the methodology of Heukeshoven and Dernick, the kit gives essentially colorless backgrounds in most gel electrophoresis systems.
  • Silver Staining Kit, Protein provides convenient, reproducible and sensitive staining of proteins in polyacrylamide gels .including PhastGels in less than two hours
  • The visualization technique detects most proteins in the nanogram range, which is 100-fold more sensitive than Coomassie Blue staining.
  • Typical sensitivity is 0.2 to 0.6 ng protein/band.
  • Use with 2-D electrophoresis and mass spectrometry by omitting glutaraldehyde from staining procedure.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Environment

Signal Word

Danger

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrián Matencio et al.
Pharmaceuticals (Basel, Switzerland), 13(9) (2020-09-03)
Our desire to live longer has led to an ever-increasing number of novel antiaging products. However, few molecules have any real effect and new ones need to be studied before they can be used commercially. In this contribution, activation of

Articles

This page shows how to perform a purification and homogeneity check of membrane proteins with products from Cytiva.

This page shows how to perform a purification of His-tagged membrane proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service