GE10600062
Amersham™ Protran® Western blotting membranes, nitrocellulose
pore size 0.45 μm, sheet W × L 140 mm × 160 mm, pkg of 25 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 25 ea
manufacturer/tradename
Cytiva 10600062
sheet W × L
140 mm × 160 mm
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Nitrocellulose (NC) membrane for use in Western blot protocols, offering low background and high signal-to-noise ratios. Amersham Protran 0.45 NC is a nitrocellulose membrane with a 0.45 µm pore size for use in Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background noise, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western Blotting detection reagents. Protran NC membranes are available in popular precut sizes for a wide range of laboratory applications. This product was formerly called Protran BA85.
Application
Amersham™ Protran® Western blotting membranes, nitrocellulose is suitable for use in:
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- Low background and excellent signal-to-noise ratios
- Compatible with a variety of labelling and detection systems
- High binding capacity
- Suitable for proteins with a wide range of molecular weights
- Ideal tool for your antibody purification and analysis workflow
- Proprietary nitrocellulose formula ensures a long shelf life of bound proteins
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service