About This Item
Recommended Products
mp
108-110 °C (lit.)
storage temp.
−20°C
SMILES string
Cl[H].CCOC(=O)CC[C@H](N)C(=O)OCC
InChI
1S/C9H17NO4.ClH/c1-3-13-8(11)6-5-7(10)9(12)14-4-2;/h7H,3-6,10H2,1-2H3;1H/t7-;/m0./s1
InChI key
WSEQLMQNPBNMSL-FJXQXJEOSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service