Skip to Content
Merck
All Photos(2)

Key Documents

G8759

Sigma-Aldrich

D-Glucose diethylmercaptal

Synonym(s):

D-Glucose diethyl dithioacetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O5S2
CAS Number:
Molecular Weight:
286.41
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

storage temp.

2-8°C

SMILES string

CCSC(SCC)C(O)C(O)C(O)C(O)CO

InChI

1S/C10H22O5S2/c1-3-16-10(17-4-2)9(15)8(14)7(13)6(12)5-11/h6-15H,3-5H2,1-2H3/t6-,7-,8+,9-/m1/s1

InChI key

BTOYCPDACQXQRS-LURQLKTLSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Iwamoto et al.
Bioscience, biotechnology, and biochemistry, 59(3), 408-411 (1995-03-01)
When D-glucosaminate dehydratase (GADH) was incubated with D-glucosaminate (GlcNA) in veronal buffer (VB; 0.01 M, pH 8.0), GlcNA was converted stoichiometrically to glyceraldehyde, pyruvate, and ammonia (aldolase reaction A). This reaction occurred in addition to the dehydratase reaction (conversion of
Ryoko Iwamoto et al.
Biochimica et biophysica acta, 1647(1-2), 310-314 (2003-04-11)
The complete amino acid sequence of the D-glucosaminate dehydratase (GADH) alpha-subunit from Pseudomonas fluorescens was determined by PCR using genomic DNA from P. fluorescens as a template. The alpha-subunit comprises 320 amino acids and has a molecular mass of about
Wei-Wei Duan et al.
Huan jing ke xue= Huanjing kexue, 33(1), 26-31 (2012-03-29)
The aim was to compare the characteristics and the differences in carbon catabolic diversity of air samples collected from five locations that around the edge of Taklamakan desert. The characteristics and the differences of carbon metabolic profiles were detected by
Sheng-Ju Ou et al.
Archiv der Pharmazie, 339(9), 527-530 (2006-08-31)
Two chromium(III) complexes of glucosaminic acid were synthesized by neutralization and exchange reaction. The formation of 1 : 1 and 2 : 3 (Cr : glucosaminate) complexes was confirmed by elemental analyses and spectroscopic studies. The effect of the complexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service