Skip to Content
Merck
All Photos(1)

Key Documents

G8511

Supelco

β-Galactosidase from Escherichia coli

For use as a marker in SDS-PAGE

Synonym(s):

β-D-Galactoside galactohydrolase, Lactase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25

mol wt

~116 kDa

packaging

vial of 0.5 mg

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

β-Galactosidase from Escherichia coli has been used as an internal standard in sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE).[1] It has been used in the protein ladder mix for optimization of microfluidic western blotting assay.[2]

Biochem/physiol Actions

β-galactosidase cleaves lactose into its monosaccharide components, glucose and galactose. It also catalyses the transglycosylation of glucose into allolactose, the inducer of β-galactosidase, in a feedback loop.

Physical properties

Tetramer molecular weight 465 kDa (subunits 116.3 kDa each)

Unit Definition

One unit will hydrolyze 1.0 μmole of o-nitrophenyl β-D-galactoside to o-nitrophenol and D-galactose per min at pH 7.3 at 37 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microfluidic Western blotting of low-molecular-mass proteins.
Gerver RE and Herr AE
Analytical Chemistry, 86, 10625-10632 (2014)
Quantitative determination of titin and nebulin in poultry meat by SDS-PAGE with an internal standard.
Tomaszewska-Gras J, et al.
Meat Science, 62, 61-66 (2002)
Scott W McPherson et al.
Journal of immunology (Baltimore, Md. : 1950), 190(10), 4971-4981 (2013-04-16)
Extrathymically derived regulatory T cells (iTregs) protect against autoimmunity to tissue-specific Ags. However, whether Ag-specific iTreg generation and function is limited to secondary lymphoid tissue or whether it can occur within the tissue-specific local environment of the cognate Ag remains
Staci Rakowiecki et al.
Development (Cambridge, England), 140(8), 1730-1739 (2013-03-15)
The morphogenetic program that shapes the three semicircular canals (SSCs) must be executed with extreme precision to satisfy their complex vestibular function. The SSCs emerge from epithelial outgrowths of the dorsal otocyst, the central regions of which fuse and resorb
Yiquan Zhang et al.
Future microbiology, 8(2), 271-280 (2013-02-05)
To explore the transcriptional regulation of the psaEF and psaABC loci by the RovA and PhoP regulators in Yersinia pestis. Primer extension, LacZ fusion, gel mobility shift and DNase I footprinting assays were conducted in combination for this gene regulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service