Skip to Content
Merck
All Photos(1)

Documents

G6758

Sigma-Aldrich

Glutaryl-L-phenylalanine 7-amido-4-methylcoumarin

Synonym(s):

Glut-Phe-AMC, N-Glutaryl-L-phenylalanine-4-methyl-7-coumarinylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H24N2O6
CAS Number:
Molecular Weight:
436.46
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

powder

fluorescence

λex 325 nm; λem 395 nm in H2O
λex 380 nm; λem 460 nm (Reaction product)

storage temp.

−20°C

SMILES string

CC1=CC(=O)Oc2cc(NC(=O)[C@H](Cc3ccccc3)NC(=O)CCCC(O)=O)ccc12

InChI

1S/C24H24N2O6/c1-15-12-23(30)32-20-14-17(10-11-18(15)20)25-24(31)19(13-16-6-3-2-4-7-16)26-21(27)8-5-9-22(28)29/h2-4,6-7,10-12,14,19H,5,8-9,13H2,1H3,(H,25,31)(H,26,27)(H,28,29)/t19-/m0/s1

InChI key

OTPFOMJAYSBYOD-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service