Skip to Content
Merck
All Photos(1)

Documents

G4384

Sigma-Aldrich

Gly-Gly-Ile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19N3O4
CAS Number:
Molecular Weight:
245.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CNC(=O)CN)C(O)=O

InChI

1S/C10H19N3O4/c1-3-6(2)9(10(16)17)13-8(15)5-12-7(14)4-11/h6,9H,3-5,11H2,1-2H3,(H,12,14)(H,13,15)(H,16,17)/t6-,9-/m0/s1

InChI key

XMPXVJIDADUOQB-RCOVLWMOSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Rohm et al.
Molecular nutrition & food research, 63(5), e1801094-e1801094 (2018-12-07)
Peptide transporter 1 (PEPT1) function is well understood, yet little is known about its contribution toward the absorption of dietary amino acids in the form of di- and tripeptides. In the present human study, changes in plasma concentrations of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service