Skip to Content
Merck
All Photos(1)

Key Documents

F8883

Sigma-Aldrich

2′-Fluoro-2′-deoxycytidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN3O4
CAS Number:
Molecular Weight:
245.21
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)C2OC(CO)C(O)C2F

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Helmling et al.
Nucleosides, nucleotides & nucleic acids, 22(5-8), 1035-1038 (2003-10-21)
Synthesis of 2'-fluoro-nucleosides from L-arabinose in order to perform the synthesis of 2'-fluoro-Spiegelmers binding to a neuropeptide.
S Schmidt et al.
Biochimica et biophysica acta, 1130(1), 41-46 (1992-02-28)
Protected 2'-deoxy-2'-fluorouridine and 2'-deoxy-2'-fluorocytidine suitable for incorporation into oligonucleotides via the phosphoramidite approach have been prepared. Five modified and two unmodified oligonucleotides have been synthesized to investigate the regiospecific cleavage of a 5S RNA from Escherichia coli by RNase H.
Robert S Jansen et al.
Rapid communications in mass spectrometry : RCM, 23(19), 3040-3050 (2009-08-26)
A novel assay for the simultaneous quantification of the widely used anticancer agent 2',2'-difluorodeoxycytidine (gemcitabine; dFdC), its deaminated metabolite 2',2'-difluorodeoxyuridine (dFdU) and their mono-, di- and triphosphates (dFdCMP, dFdCDP, dFdCTP, dFdUMP, dFdUDP and dFdUTP) in peripheral blood mononuclear cells (PBMCs)
Frank C Richardson et al.
Chemical research in toxicology, 15(7), 922-926 (2002-07-18)
Apatmers are synthesized using 2'-fluoropyrimdines in place of normal pyrmidines to stabilize them against enzymatic degradation, and thereby improve their therapeutic efficacy. Despite this stabilizing effect, the apatmers can still be degraded by nucleases in the blood. Primer template extension
Lieven J Stuyver et al.
Antimicrobial agents and chemotherapy, 48(2), 651-654 (2004-01-27)
2'-Deoxy-2'-fluorocytidine (FdC) is a potent inhibitor of the hepatitis C virus RNA replicon in culture, and FdC-5'-triphosphate is an effective inhibitor of the NS5B polymerase. Dynamic profiling of cell growth in an antiviral assay showed that FdC caused cytostasis due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service